missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTLA-4 Monoclonal Antibody (WKH 203), FITC
GREENER_CHOICE

Mouse Monoclonal Antibody

€ 276.00 - € 540.00

Specifications

Antigen CTLA-4
Clone WKH 203
Concentration 0.1 mg/mL
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Applications Flow Cytometry
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
15328490
GREENER_CHOICE
View Documents
Invitrogen™
MA517584
100 μg
€ 276.00
100µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
15338490
View Documents
Invitrogen™
MA517585
500 μg
€ 540.00
500µg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CTLA-4 Monoclonal Antibody for Flow

CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity than CD28. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhbits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.
Specifications

Specifications

CTLA-4
0.1 mg/mL
Flow Cytometry
FITC
Mouse
RUO
PBS with 4-5mg/mL BSA and 0.02% sodium azide
0
63835
Purified rCTLA-4hlg fusion protein.
Antibody
WKH 203
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Monoclonal
Liquid
IgG1
Rat
CTLA4
ALPS5; CD; CD152; CD152 antigen; CD152 isoform; CD152 protein; CD152 protein precursor; celiac disease 3; CELIAC3; Ctla4; CTLA-4; cytotoxic T lymphocyte associated antigen 4 short spliced form; cytotoxic T-lymphocyte associated protein 4; cytotoxic T-lymphocyte protein 4; cytotoxic T-lymphocyte protein 4 isoform CTLA4-TM; cytotoxic T-lymphocyte-associated antigen 4; cytotoxic T-lymphocyte-associated protein 4; cytotoxic T-lymphocyte-associated serine esterase-4; EGK_04712; GRD4; GSE; ICOS; IDDM12; insulin-dependent diabetes mellitus 12; ligand and transmembrane spliced cytotoxic T lymphocyte associated antigen 4; Ly-56; sCTLA4; soluble form; transmembrane form
CTLA4
Primary
Protein G
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.