missing translation for 'onlineSavingsMsg'
Learn More

Abnova™ CTLA4 (Human) Recombinant Protein

Product Code. 16083875
Click to view available options
Quantity:
2 μg
Unit Size:
2µg
This item is not returnable. View return policy

Product Code. 16083875

Brand: Abnova™ H00001493H01.2ug

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Purified CTLA4 (AAH74842.1, 37 a.a. - 161 a.a.) human recombinant protein with His-Flag-StrepII tag at N-terminus expressed in human cells.

  • Sequence: AMHVAQPAVVLASSRGIASFVCEYASPGKATEVRVTVLRQADSQVTEVCAATYMMGNELTFLDDSICTGTSSGNQVNLTIQGLRAMDTGLYICKVELMYPPPYYLGIGNGTQIYVIDPEPCPDSD

Specifications

Accession Number AAH74842.1
Concentration ≥ 10 μg/ml
Gene ID (Entrez) 1493
Name cytotoxic T-lymphocyte-associated protein 4
Preparation Method Transfection of pSuper-CTLA4 plasmid into HEK293T cell, and the expressed protein was purified by Strep-Tactin affinity column
Quality Control Testing SDS-PAGE and Western Blot
Quantity 2 μg
Storage Requirements Store at -80°C. Aliquot to avoid repeated freezing and thawing.
Gene Alias CD152, CELIAC3, CTLA-4, GSE, IDDM12
Cell Type Human HEK293T cells
Gene Symbol CTLA4
Species Human
Protein Tag His-Flag-StrepII
Buffer 100 mM Tris-HCl pH 8.0, 150 mM NaCl, 1 mM EDTA, and 5 mM desthiobiotin
Form Liquid
Show More Show Less
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.