Learn More
SDC1 Monoclonal Antibody (OTI2F8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA813758S
Description
Dilute in PBS (pH7.3) if necessary. Stable for 12 months from date of receipt.
SDC1 Monoclonal antibody specifically detects SDC1 in Human samples. It is validated for Immunohistochemistry (Paraffin)Specifications
| SDC1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P18827 | |
| Mouse | |
| Protein A/G | |
| RUO | |
| 6382 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| OTI2F8 | |
| Unconjugated | |
| Sdc1 | |
| AA408134; AA409076; CD antigen 138; CD138; CD138 antigen; heparan sulfate proteoglycan fibroblast growth factor receptor; HSPG; sCD138; SDC; Sdc1; soluble CD138; Sstn; syn-1; Synd; SYND1; Synd-1; SYNDECA; syndecan; syndecan 1; syndecan proteoglycan 1; Syndecan1; syndecan-1; synstatin | |
| Full length human recombinant protein of human SDC1 (NP_001006947) produced in HEK293T cell. | |
| 30 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.