Learn More
CTLA4 Mouse anti-Human, Clone: OTI8D12, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF807714
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
CTLA4 (Cytotoxic T-Lymphocyte Antigen 4) is a CD28-family receptor expressed on mainly CD4+ T cells. It binds the same ligands as CD28 (CD80 and CD86 on B cells and dendritic cells), but with higher affinity. However, in contrast to CD28 which enhances cell function when bound at the same time as the T cell receptor, CTLA4 inhibits the T cell and prevents it from functioning. Intracellular CTLA4 is also found in regulatory T cells and may be important to their function.Specifications
| CTLA4 | |
| Monoclonal | |
| Unconjugated | |
| CTLA4 | |
| ALPS5, CD, CD152, CELIAC3, CTLA-4, GRD4, GSE, IDDM12 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1493 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Western Blot | |
| OTI8D12 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| P16410 | |
| CTLA4 | |
| Full length human recombit protein of human CTLA4 produced in HEK293T cell. | |
| 100 μg | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.