missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTNND1 Monoclonal Antibody (OTI1G4), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA800968
This item is not returnable.
View return policy
Description
Delta 1 Catenin is an efficient tyrosine kinase substrate implicated both in cell transformation by SRC and in ligand-induced receptor signaling through the EGF, PDGF, CSF-1 and ERBB2 receptors. The association of catenins to cadherins produces a complex which is linked to the actin filament network, and which seems to be of primary importance for cadherins cell-adhesion properties.Specifications
| CTNND1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| O60716 | |
| CTNND1 | |
| Full length human recombit protein of human CTNND1 produced in HEK293T cell. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1G4 | |
| Unconjugated | |
| CTNND1 | |
| BCDS2, CAS, CTNND, P120CAS, P120CTN, p120, p120(CAS), p120(CTN) | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 1500 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction