missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ CTR9 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA530323

Product Code. 13454817

  • € 520.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Recommended positive controls: 293T, A431, HeLa, HepG2. Predicted reactivity: Mouse (100%), Rat (100%), Zebrafish (95%), Xenopus laevis (97%), Bovine (100%). Store product as a concentrated solution. Centrifuge briefly prior to opening the vial.

MED18 is a component of the Mediator complex, which is a coactivator for DNA-binding factors that activate transcription via RNA polymerase II.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

CTR9
Polyclonal
Unconjugated
Ctr9
AA409336; Ctr9; CTR9 homolog, Paf1/RNA polymerase II complex component; CTR9, Paf1/RNA polymerase II complex component; Ctr9, Paf1/RNA polymerase II complex component, homolog; Ctr9, Paf1/RNA polymerase II complex component, homolog (S. cerevisiae); Kiaa0155; mKIAA0155; p150; p150TSP; RNA polymerase-associated protein CTR9 homolog; SH2 domain binding protein 1 (tetratricopeptide repeat containing); SH2 domain-binding protein 1; SH2BP1; Tetratricopeptide repeat-containing, SH2-binding phosphoprotein of 150 kDa; TPR-containing, SH2-binding phosphoprotein; TPR-containing, SH2-binding phosphoprotein of 150 kDa; TSBP; Tsp
Rabbit
Antigen affinity chromatography
RUO
9646
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
1.03 mg/mL
PBS with 1% BSA, 20% glycerol and 0.01% thimerosal; pH 7
Q6PD62
Ctr9
Recombinant protein encompassing a sequence within the N-terminus region of human CTR9. The exact sequence is proprietary.
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.