missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP2 Antibody (1D7-2C7), Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals H00114898-M01
This item is not returnable.
View return policy
Description
CTRP2 Monoclonal antibody specifically detects CTRP2 in Human samples. It is validated for Western Blot, ELISA
Specifications
| CTRP2 | |
| Monoclonal | |
| Unconjugated | |
| In 1x PBS, pH 7.4 | |
| C1q and tumor necrosis factor related protein 2, complement-c1q tumor necrosis factor-related protein 2, CTRP2complement C1q tumor necrosis factor-related protein 2, zacrp2 | |
| C1QTNF2 (AAH11699.1, 16 a.a. ∽ 285 a.a) full-length recombinant protein with GST tag. MW of the GST tag alone is 26 KDa. DPLLGAFARRDFRKGSPQLVCSLPGPQGPPGPPGAPGPSGMMGRMGFPGKDGQDGHDGDRGDSGEEGPPGRTGNRGKPGPKGKAGAIGRAGPRGPKGVNGTPGKHGTPGKKGPKGKKGEPGLPGPCSCGSGHTKSAFSVAVTKSYPRERLPIKFDKILMNEGGHYNASSGKFVCGVPGIYYFTYDITLANKHLAIGLVHNGQYRIRTFDANTGNHDVASGSTILALKQGDEVWLQIFYSEQNGLFYDPYWTDSLFTGFLIYADQDDPNEV | |
| 0.1 mg | |
| Signal Transduction | |
| 114898 | |
| Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles. | |
| IgG2a κ |
| Western Blot, ELISA | |
| 1D7-2C7 | |
| Western Blot 1:500, ELISA | |
| AAH11699 | |
| Mouse | |
| IgG purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction