missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP2 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76627UV
This item is not returnable.
View return policy
Description
CTRP2 Polyclonal antibody specifically detects CTRP2 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| CTRP2 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| DyLight 350 | |
| C1q and tumor necrosis factor related protein 2, complement-c1q tumor necrosis factor-related protein 2, CTRP2complement C1q tumor necrosis factor-related protein 2, zacrp2 | |
| Antibody was raised against a 16 amino acid synthetic peptide from near the center of human CTRP2. The immunogen is located within amino acids 140 - 190 of CTRP2. Amino Acid Squence: VTKSYPRERLPIKFDK | |
| 0.1 mL | |
| Signal Transduction | |
| 114898 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction