missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTRP4 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559403
This item is not returnable.
View return policy
Description
Immunogen sequence: GPGSSELRSA FSAARTTPLE GTSEMAVTFD KVYVNIGGDF DVATGQFRCR VPGAYFFSFT Highest antigen sequence identity to the following orthologs: Mouse - 97%, Rat - 97%.
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. Multiple isoforms of mouse CTRP4 have been reported.
Specifications
| CTRP4 | |
| Polyclonal | |
| Unconjugated | |
| C1QTNF4 | |
| 0710001E10Rik; 9430004J15Rik; C1q and tumor necrosis factor related protein 4; C1q/TNF-related protein 4; C1QTNF4; Complement C1q tumor necrosis factor-related protein 4; complement-c1q tumor necrosis factor-related protein 4; Ctrp4; ZACRP4 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114900 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BXJ3 | |
| C1QTNF4 | |
| Recombinant protein corresponding to Human CTRP4. Recombinant protein control fragment (Product #RP-97469). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction