missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTRP5 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA520152
This item is not returnable.
View return policy
Description
A suggested positive control is human brain tissue lysate. PA5-20152 can be used with blocking peptide PEP-0272.
Adipose tissue of an organism plays a major role in regulating physiologic and pathologic processes such as metabolism and immunity by producing and secreting a variety of bioactive molecules termed adipokines. One highly conserved family of adipokines is adiponectin/ACRP30 and its structural and functional paralogs, the C1q/tumor necrosis factor-alpha-related proteins (CTRPs) 1-7. Unlike adiponectin, which is expressed exclusively by differentiated adipocytes, the CTRPs are expressed in a wide variety of tissues. These proteins are thought to act mainly on liver and muscle tissue to control glucose and lipid metabolism. An analysis of the crystal structure of adiponectin revealed a structural and evolutionary link between TNF and C1q-containing proteins, suggesting that these proteins arose from a common ancestral innate immunity gene. CTRP5 has been suggested to be involved in age-related macular degeneration.
Specifications
| CTRP5 | |
| Polyclonal | |
| Unconjugated | |
| C1QTNF5 | |
| C1q and tumor necrosis factor related protein 5; C1q TNF-alpha-related protein 5; C1qtnf5; Complement C1q tumor necrosis factor-related protein 5; Ctrp5; LORD; Membrane frizzled-related protein; Membrane-type frizzled-related protein; Mfrp; myonectin; UNQ303/PRO344 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 114902, 315598 | |
| Maintain refrigerated at 2-8°C for up to 3 months. For long term storage store at -20°C | |
| Liquid |
| Immunohistochemistry, Western Blot | |
| 1 mg/mL | |
| PBS with 0.02% sodium azide | |
| Q5FVH0, Q9BXJ0 | |
| C1QTNF5 | |
| A 15 amino acid peptide from near the amino terminus of human CTRP5. | |
| 100 μg | |
| Primary | |
| Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction