missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTRP6 Antibody, DyLight 350, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-77218UV
This item is not returnable.
View return policy
Description
CTRP6 Polyclonal antibody specifically detects CTRP6 in Human, Mouse samples. It is validated for Western Blot, ELISA, Immunofluorescence
Specifications
| CTRP6 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
| Western Blot, ELISA, Immunofluorescence | |
| DyLight 350 | |
| C1q and tumor necrosis factor related protein 6, complement C1q tumor necrosis factor-related protein 6, complement-c1q tumor necrosis factor-related protein 6, CTRP6ZACRP6 | |
| Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CTRP6. The immunogen is located within the last 50 amino acids of CTRP6. Amino Acid Squence: RLFKRQRENAIYSND | |
| 0.1 mL | |
| Signal Transduction | |
| 114904 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction