Learn More
CTRP7 Antibody, PerCP, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand:  Novus Biologicals NBP1-76634PCP
Product Code. 18572354
  • € 564.00 / 0.10mL
Description
CTRP7 Polyclonal antibody specifically detects CTRP7 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
TRUSTED_SUSTAINABILITY
Specifications
Specifications
CTRP7
Polyclonal
Western Blot, ELISA, Immunocytochemistry/ Immunofluorescence
C1q and tumor necrosis factor related protein 7, complement-c1q tumor necrosis factor-related protein 7, CTRP7complement C1q tumor necrosis factor-related protein 7, ZACRP7
Antibody was raised against a 15 amino acid synthetic peptide from near the carboxy terminus of human CTRP7. The immunogen is located within the last 50 amino acids of CTRP7. Amino Acid Squence: YVDTDYLDSISEDDE
0.1 mL
Chemokines and Cytokines
114905
Store at 4C in the dark.
IgG
Western Blot, ELISA, Immunocytochemistry
PerCP
PBS
Rabbit
Peptide affinity purified
RUO
Primary
Human, Mouse, Rat