missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTSL1 Monoclonal Antibody (OTI5C8), TrueMAB™, OriGene
Mouse Monoclonal Antibody
Brand: Origene Technologies TA812846S
This item is not returnable.
View return policy
Description
CTSL1 Monoclonal antibody specifically detects CTSL1 in Human samples. It is validated for Immunohistochemistry (Paraffin), Western BlotSpecifications
| CTSL1 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide | |
| P07711 | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 1514 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI5C8 | |
| Unconjugated | |
| CTSL | |
| 1190035F06Rik; Cat L; cathepsin L; Cathepsin L1; Cathepsin L1 heavy chain; Cathepsin L1 light chain; CATHL; CatL; CP-2; Ctsl; CTSL1; cyclic protein 2; fs; furless; major excreted protein; MEP; nackt; nkt; p39 cysteine proteinase; Procathepsin L | |
| Human recombinant protein fragment corresponding to amino acids 114-333 of human CTSL produced in E.coli. | |
| 30 μL | |
| Primary | |
| Human | |
| Liquid |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction