missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CTU1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101194
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total ATPBD3.
Plays a central role in 2-thiolation of mcm(5)S(2)U at tRNA wobble positions of tRNA(Lys), tRNA(Glu) and tRNA(Gln). Directly binds tRNAs and probably acts by catalyzing adenylation of tRNAs, an intermediate required for 2-thiolation. It is unclear whether it acts as a sulfurtransferase that transfers sulfur from thiocarboxylated URM1 onto the uridine of tRNAs at wobble position.
Specifications
| CTU1 | |
| Polyclonal | |
| Unconjugated | |
| CTU1 | |
| ATP binding domain 3; Atpbd3; ATP-binding domain-containing protein 3; BC005752; Cancer-associated gene protein; cancer-associated protein; CTU1; Cytoplasmic tRNA 2-thiolation protein 1; cytoplasmic tRNA adenylyltransferase 1; cytosolic thiouridylase subunit 1; cytosolic thiouridylase subunit 1 homolog; cytosolic thiouridylase subunit 1 homolog (S. pombe); NCS6 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 90353 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q7Z7A3 | |
| CTU1 | |
| A synthesized peptide derived from human CTU1(Accession Q7Z7A3), corresponding to amino acid residues L298-F348. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction