missing translation for 'onlineSavingsMsg'
Learn More

CTX-0294885 hydrochloride, MedChemExpress

Product Code. 16426958
Encompass_Preferred
Change view
Click to view available options
Quantity:
5 mg
10 mg
50 mg
100 mg
Unit Size:
100mg
10mg
50mg
5mg
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity Unit Size
16426958 10 mg 10mg
16493007 50 mg 50mg
16469497 5 mg 5mg
16407036 100 mg 100mg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
This item is not returnable. View return policy
Product Code. 16426958 Supplier Medchem Express Supplier No. HY15985A
 Restricted Item
Estimated Shipment: 06-10-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

MedChemExpress CTX-0294885 hydrochloride is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 hydrochloride is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.

CTX-0294885 hydrochloride is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 hydrochloride is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.

Specifications

Chemical Name or Material CTX-0294885 hydrochloride
Physical Form Solid
Color Off-White
Quantity 10 mg
Purity Grade Notes Research
Molecular Formula C22H25Cl2N7O
Solubility Information H2O : 55 mg/mL (115.94 mM; Need ultrasonic) ∣DMSO : 9.17 mg/mL (19.33 mM; Need ultrasonic)
SMILES [H]Cl.O=C(NC)C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCNCC4)C=C3)=NC=C2Cl
Molecular Weight (g/mol) 474.39
Formula Weight 474.39
Percent Purity 99.93%
Grade Research
Show More Show Less

Research purposes only

Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.