missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CTX-0294885 hydrochloride, MedChemExpress
Description
CTX-0294885 hydrochloride is a broad spectrum kinase inhibitor that can capture 235 kinases from MDA-MB-231 cells, and can capture all members of the AKT family. CTX-0294885 hydrochloride is a powerful reagent for analysis of kinome signaling networks that can be used for the research of diseases like inflammation, diabetes, and cancer.
Specifications
Specifications
| Chemical Name or Material | CTX-0294885 hydrochloride |
| Physical Form | Solid |
| Color | Off-White |
| Quantity | 50 mg |
| Purity Grade Notes | Research |
| Molecular Formula | C22H25Cl2N7O |
| Solubility Information | H2O : 55 mg/mL (115.94 mM; Need ultrasonic) ∣DMSO : 9.17 mg/mL (19.33 mM; Need ultrasonic) |
| SMILES | [H]Cl.O=C(NC)C1=CC=CC=C1NC2=NC(NC3=CC=C(N4CCNCC4)C=C3)=NC=C2Cl |
| Molecular Weight (g/mol) | 474.39 |
| Formula Weight | 474.39 |
| Show More |
Research purposes only
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?