missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ Cubilin Antibody (SR2571), Novus Biologicals™
Rabbit Monoclonal Antibody
€ 339.00 - € 406.00
Specifications
| Antigen | Cubilin |
|---|---|
| Clone | SR2571 |
| Dilution | Western Blot 1:500-1:2000 |
| Applications | Western Blot |
| Classification | Monoclonal |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
30041705
|
Novus Biologicals™
NBP3-27698-50ul |
50 μL |
€ 339.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
30041572
|
Novus Biologicals™
NBP3-27698-100ul |
100 μL |
€ 406.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Cubilin Monoclonal antibody specifically detects Cubilin in Human,Mouse,Rat samples. It is validated for Western BlotSpecifications
| Cubilin | |
| Western Blot 1:500-1:2000 | |
| Monoclonal | |
| Purified | |
| RUO | |
| PBS (pH 7.4), 150mM NaCl, 50% glycerol | |
| 8029 | |
| IgG | |
| Affinity purified |
| SR2571 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse, Rat | |
| cubilin, cubilin (intrinsic factor-cobalamin receptor), gp280, IFCR, MGA1 | |
| A synthesized peptide derived from human Cubilin (Uniprot #: O60494) | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze/thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title