missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC1 Antibody, Alexa Fluor™ 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-76946AF594
This item is not returnable.
View return policy
Description
CUEDC1 Polyclonal antibody specifically detects CUEDC1 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
| CUEDC1 | |
| Polyclonal | |
| 50mM Sodium Borate | |
| Rabbit | |
| Peptide affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse, Rat | |
| Purified |
| Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 594 | |
| CUE domain containing 1, CUE domain-containing protein 1, DKFZp547L163, FLJ20739 | |
| Antibody was raised against a 16 amino acid synthetic peptide near the carboxy terminus of human CUEDC1. The immunogen is located within amino acids 310 - 360 of CUEDC1. Amino Acid Squence: AFSEKTKMRKSKRKHL | |
| 0.1 mL | |
| Cancer | |
| 404093 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction