missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC1 Rabbit anti-Human, Rat, Polyclonal, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 178.00 - € 383.00
Specifications
| Antigen | CUEDC1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Form | Purified |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18326715
|
Novus Biologicals
NBP3-16054-20UL |
20 μg |
€ 178.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18389723
|
Bio-Techne
NBP3-16054-100UL |
100 μg |
€ 383.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CUEDC1 Polyclonal antibody specifically detects CUEDC1 in Human, Rat samples. It is validated for Western BlotSpecifications
| CUEDC1 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human, Rat | |
| CUE domain containing 1, CUE domain-containing protein 1, DKFZp547L163, FLJ20739 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 187-386 of human CUEDC1 (NP_001258804.1). | |
| Primary | |
| Store at -20°C. Avoid freeze-thaw cycles. |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Cancer | |
| PBS, 50% glycerol, pH7.3 | |
| 404093 | |
| IgG | |
| Affinity purified |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title