missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-24684SS
This item is not returnable.
View return policy
Description
CUEDC2 Polyclonal specifically detects CUEDC2 in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin.
Specifications
| CUEDC2 | |
| Polyclonal | |
| Unconjugated | |
| 0.2 ml PBS and 0.05% BSA with 0.05% Sodium Azide | |
| bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| 79004 | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| 0.5 mg/mL | |
| Immunohistochemistry-Paraffin 5 ug/ml | |
| Q9H467 | |
| CUEDC2 | |
| A portion of amino acid 50-100 of human CUEDC2 was used as the immunogen. | |
| 0.025 mg | |
| Primary | |
| The amino acid sequence used as immunogen is 100% homologous in human, rhesus monkey, chimpanzee, cow, rat, mouse, wild boar, horse, opossum, zebrafinch, platypus, rabbit and Xenopus and 94% homologous in zebrafish and pufferfish. | |
| Store at 4C short term. Aliquot and store at -20C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction