missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUEDC2 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
€ 216.00 - € 326.00
Specifications
| Antigen | CUEDC2 |
|---|---|
| Dilution | Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 |
| Applications | Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
18683188
|
Novus Biologicals
NBP2-99801-100ul |
100 μg |
€ 326.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
18613098
|
Novus Biologicals
NBP2-99801-50ul |
50 μg |
€ 216.00
50µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
CUEDC2 Polyclonal antibody specifically detects CUEDC2 in Human samples. It is validated for Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin)Specifications
| CUEDC2 | |
| Western Blot, ELISA, Immunoprecipitation, Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| Rabbit | |
| Signal Transduction | |
| 0.2 um filtered solution in PBS | |
| 79004 | |
| IgG | |
| Antigen and protein A Affinity-purified |
| Western Blot 1:500-1:1000, ELISA 1:25000-1:50000, Immunoprecipitation 1-2 uL/mg of lysate, Immunohistochemistry-Paraffin 1:2000-1:10000 | |
| Polyclonal | |
| Purified | |
| RUO | |
| Human | |
| bA18I14.5, C10orf66, chromosome 10 open reading frame 66, CUE domain containing 2, CUE domain-containing protein 2, MGC2491 | |
| Produced in rabbits immunized with purified, recombinant Human CUEDC2. | |
| Primary | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title