missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUGBP1/CELF1 Antibody (OTI5B8), Alexa Fluor™ 532, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-71623AF532
This item is not returnable.
View return policy
Description
CUGBP1/CELF1 Monoclonal specifically detects CUGBP1/CELF1 in Human samples. It is validated for Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence.
Specifications
| CUGBP1/CELF1 | |
| Monoclonal | |
| Alexa Fluor 532 | |
| 50 mM sodium borate with 0.05% sodium azide | |
| CELF1 | |
| Full length recombinant protein of human CUGBP1/CELF1 (NP_006551) produced in HEK293T cells. | |
| Primary | |
| Human | |
| IgG2b |
| Western Blot, Immunohistochemistry, Immunocytochemistry, Immunofluorescence | |
| OTI5B8 | |
| Western Blot, Immunohistochemistry, Immunocytochemistry/Immunofluorescence | |
| BRUNOL250 kDa nuclear polyadenylated RNA-binding protein, bruno-like 2, Bruno-like protein 2, CUG RNA-binding protein, CUG triplet repeat, RNA binding protein 1, CUG triplet repeat, RNA-binding protein 1, CUG-BP, CUG-BP- and ETR-3-like factor 1, CUGBP Elav-like family member 1, CUGBP, Elav-like family member 1, CUGBP1CUG triplet repeat RNA-binding protein 1, CUGBPCELF-1, Deadenylation factor CUG-BP, EDEN-BP, EDEN-BP homolog, embryo deadenylation element binding protein, Embryo deadenylation element-binding protein homolog, hNab50, NAB50CUG-BP1, NAPOR, nuclear polyadenylated RNA-binding protein, 50-kD, RNA-binding protein BRUNOL-2 | |
| Mouse | |
| 0.1 mL | |
| 10658 | |
| Store at 4°C in the dark. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction