Learn More
CUL2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A6G9, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-67175-100UL
Description
The CUL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL2. This antibody recognizes human, rat antigen. The CUL2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
| CUL2 | |
| 1A6G9 | |
| CoraLite Plus 488 | |
| Mouse | |
| Primary | |
| Human, Rat | |
| 155 kDa | |
| IgG1 |
| Immunofluorescence | |
| 1000 μg/mL | |
| CUL 2, CUL2, cullin 2 | |
| 100 μL | |
| 8453 | |
| Store at -20°C. Avoid exposure to light. Stable for one year after shipment. | |
| Liquid |