Learn More
CUL2 Mouse, anti-Human, CoraLite Plus 488, Clone: 1A6G9, Proteintech
Mouse Monoclonal Antibody
Brand:  Proteintech CL488-67175-100UL
 View more versions of this product
Product Code. 18090636
  • € 450.00 / 100µL
Description
The CUL2 antibody from Proteintech is a mouse monoclonal antibody generated with recombinant protein of human CUL2. This antibody recognizes human, rat antigen. The CUL2 antibody has been validated for the following applications: IF analysis. This antibody is coralite™ plus 488.
Specifications
Specifications
CUL2
1A6G9
CoraLite Plus 488
Mouse
Primary
Human, Rat
155 kDa
IgG1
Immunofluorescence
1000 μg/mL
CUL 2, CUL2, cullin 2
100 μL
8453
Store at -20°C. Avoid exposure to light. Stable for one year after shipment.