missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CUL4B Antibody, Alexa Fluor™ 750, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-97399AF750
This item is not returnable.
View return policy
Description
CUL4B Polyclonal specifically detects CUL4B in Human samples. It is validated for Immunocytochemistry/Immunofluorescence.
Specifications
| CUL4B | |
| Polyclonal | |
| Immunocytochemistry/Immunofluorescence | |
| CUL-4B, cullin 4B, cullin-4B, DKFZp686F1470, KIAA0695, MRXHF2, MRXS15, MRXSC, SFM2 | |
| Produced in rabbits immunized with E. coli-derived Human CUL4B fragment. | |
| 0.1 mL | |
| Cell Cycle and Replication | |
| 8450 | |
| Store at 4°C in the dark. | |
| IgG |
| Immunofluorescence | |
| Alexa Fluor 750 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Antigen Affinity-purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction