Learn More
CUL4B Mouse anti-Canine, Human, Mouse, Clone: OTI1C4, lyophilized, TrueMAB™
Mouse Monoclonal Antibody
Brand: Origene Technologies CF800444
Description
Reconstitute with PBS (pH 7.3) and recommend to perform another round of desalting process using Product No. 7KMWCO
Core component of multiple cullin-RING-based E3 ubiquitin-protein ligase complexes which mediate the ubiquitination and subsequent proteasomal degradation of target proteins. The functional specificity of the E3 ubiquitin-protein ligase complex depends on the variable substrate recognition subunit. CUL4B may act within the complex as a scaffold protein, contributing to catalysis through positioning of the substrate and the ubiquitin-conjugating enzyme. Plays a role as part of the E3 ubiquitin-protein ligase complex in polyubiquitination of CDT1, histone H2A, histone H3 and histone H4 in response to radiation-induced DNA damage. Targeted to UV damaged chromatin by DDB2 and may be important for DNA repair and DNA replication. Required for ubiquitination of cyclin E, and consequently, normal G1 cell cycle progression. Regulates the mammalian target-of-rapamycin (mTOR) pathway involved in control of cell growth, size and metabolism. Specific CUL4B regulation of the mTORC1-mediated pathway is dependent upon 26S proteasome function and requires interaction between CUL4B and MLST8.Specifications
| CUL4B | |
| Monoclonal | |
| Unconjugated | |
| CUL4B | |
| CUL-4B, MRXHF2, MRXS15, MRXSC, SFM2 | |
| Mouse | |
| Affinity Chromatography | |
| RUO | |
| 492102, 72584, 8450 | |
| -20° C, Avoid Freeze/Thaw Cycles | |
| Lyophilized |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI1C4 | |
| PBS with 8% trehalose and no preservative; pH 7.3 | |
| A2A432, Q13620 | |
| CUL4B | |
| Human recombit protein fragment corresponding to amino acids 1-300 of human CUL4B produced in E.coli. | |
| 100 μg | |
| Primary | |
| Canine, Human, Mouse | |
| Antibody | |
| IgG2b |
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.