missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ Cullin 1 Polyclonal Antibody
GREENER_CHOICE

Product Code. 17702194
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
17702194 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 17702194 Supplier Invitrogen™ Supplier No. PA5120792

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Positive test controls include: A-549, MCF7, NIH/3T3, K-562, C6, Mouse liver, Mouse brain, Rat testis. Immunogen sequence: KNPEEAELED TLNQVMVVFK YIEDKDVFQK FYAKMLAKRL VHQNSASDDA EASMISKLKQ ACGFEYTSKL QRMFQDIGVS KDLNEQFKKH LTNSEPLDLD FSIQVLSSGS WPFQQSCTFA LPSELERSYQ RFTAFYASRH SGRKLTWLYQ LSKGELVTN.

CUL1 gene ontology annotations related to this gene include animal organ morphogenesis; apoptotic process; cell proliferation; protein monoubiquitination; SCF-dependent proteasomal ubiquitin-dependent protein catabolic process.
TRUSTED_SUSTAINABILITY

Specifications

Antigen Cullin 1
Applications ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
Classification Polyclonal
Concentration 0.88 mg/mL
Conjugate Unconjugated
Formulation PBS with 50% glycerol and 0.02% sodium azide; pH 7.3
Gene CUL1
Gene Accession No. Q13616, Q9WTX6
Gene Alias Cul1; CUL-1; cullin 1; cullin-1
Gene Symbols CUL1
Host Species Rabbit
Immunogen Recombinant fusion protein containing a sequence corresponding to amino acids 435-593 of human Cullin 1 (NP_0035832).
Purification Method Affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 26965, 362356, 8454
Target Species Human, Mouse, Rat
Content And Storage -20°C, Avoid Freeze/Thaw Cycles
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.