missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin 4a Antibody, Alexa Fluor™ 488, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-2267AF488
This item is not returnable.
View return policy
Description
Cullin 4a Polyclonal antibody specifically detects Cullin 4a in Human, Mouse samples. It is validated for Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin)
Specifications
| Cullin 4a | |
| Polyclonal | |
| Western Blot, Immunohistochemistry, Immunohistochemistry-Paraffin | |
| CUL-4A, cullin 4A, cullin-4A | |
| The immunogen recognized by this antibody maps to a region between residue 700 and the C-terminus (residue 759) of human Cullin 4A using the numbering given in entry NP_001008895.1 (GeneID 8451). | |
| 0.1 mL | |
| Apoptosis, Cancer, Tumor Suppressors | |
| 8451 | |
| Store at 4C in the dark. | |
| IgG |
| Western Blot, Immunohistochemistry, Immunohistochemistry (Paraffin) | |
| Alexa Fluor 488 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction