missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Cullin-7 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NB100-602
This item is not returnable.
View return policy
Description
Cullin-7 Polyclonal specifically detects Cullin-7 in Human, Mouse samples. It is validated for Immunoprecipitation, Western Blot (Negative).
Specifications
| Cullin-7 | |
| Polyclonal | |
| Unconjugated | |
| Tris-Citrate/Phosphate (pH 7.0 - 8.0) with 0.09% Sodium Azide | |
| CUL-7, cullin 7, cullin-7, KIAA0076dJ20C7.5 | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| Primary | |
| Human, Mouse | |
| IgG |
| Immunoprecipitation | |
| 1.0 mg/mL | |
| Immunoprecipitation 1-5 ug/mg lysate | |
| Q14999 | |
| CUL7 | |
| A synthetic peptide, mapping to a region between residue 1650 and the C-terminus (residue 1698) of human Cullin homolog 7 using the numbering given in SwissProt entry Q14999 (GeneID 9820). | |
| 100 μL | |
| Angiogenesis, DNA Repair | |
| 9820 | |
| Store at 4C. Do not freeze. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction