Learn More
R&D Systems™ Cultrex RGF Basement Membrane Extract
Product Code. 18421610
Click to view available options
Quantity:
1 mL
5 mL
2 x 5 mL
Unit Size:
10mL
1mL
5mL
Product Code. 18421610
Brand: R&D Systems™ 343300501
Applications: Bioactivity
Functional Assay - Tube formation assay: BME promotes formation of capillary-like structures by human (HBMVEC; HUVEC) or mouse (SVEC4-10) endothelial cells. Gelling Assay - BME gels in less than 30 minutes at 37degreesC, and maintains the gelled form in culture medium for a minimum of 14 days at 37degreesC.
TRUSTED_SUSTAINABILITY
Specifications
Concentration 12 to 18 mg/mL
Quantity 5 mL
Endotoxin Level ≤8EU/mL
Source Murine Engelbreth-Holm-Swarm tumor
Content And Storage Product is stable for a minimum of 3 months from date of shipment when stored at -20° C in a manual defrost freezer. For optimal stability, store at -80° C. Avoid freeze-thaw cycles.