Learn More
Greiner Bio-One Culture Tube with Two-Position Vent Cap
Polystyrene is ideally suited for optical measurements as a result of its high clarity
Brand: Greiner Bio-One 191180
Additional Details : Weight : 0.80000kg
Description
SterileSpecifications
Free of detectable DNase/RNase, human DNA and pyrogens | |
LDPE | |
Polystyrene | |
Sterile | |
Vent Stopper |
Translucent | |
Clear | |
25; 1000 | |
Not Autoclavable |