missing translation for 'onlineSavingsMsg'
Learn More

Carbolite™ Cupellation Furnace

Product Code. 15366725
Change view
Click to view available options
Capacity (Metric):
24 (no. 8 cupels)
Unit Size:
Each
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Capacity (Metric) Unit Size
15366725 24 (no. 8 cupels) Each
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15366725 Supplier Carbolite™ Supplier No. CF24230SN+&02301
  • ONLINE EXCLUSIVE PRICE
    € 46961.25
    € 24090.00 / Pack of 1
    Save € 22871.25
    48% Off
 Restricted Item
Estimated Shipment: 11-12-2026
Add to basket
Add to basket

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

Used for assay of precious metals.

  • Designed to handle the vapors without exposing the operator to hazards
  • Ensures protection from the corrosive environment which would damage conventional furnace
  • Maximum operating temperature: 1200°C
  • Variable airflow is preheated before entering the work chamber
  • Silicon carbide heating elements protected by silicon carbide tiles
  • Fumes are extracted through an insulated exhaust duct
  • Removable container collects condensed lead

Specifications

Capacity (Metric) 24 (no. 8 cupels)
For Use With (Equipment) Assay of precious metals
Temperature (Metric) 1200°C
Type Cupellation Furnace
Style Thermocouple Type: R
Dimensions (D x W x H) Interior 340 x 250 x 200 mm
Dimensions (L x W x H) Exterior W x H: 1000 x 2050 mm
Depth (Metric) 1200 mm
Wattage 13000 w
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.