missing translation for 'onlineSavingsMsg'
Learn More

Custom PreAmp Pools

Product Code. 15504364 Shop All Applied Biosystems Products
Click to view available options
Format:
96A
96B
Format 12
Format 16
Format 24
Format 32
Format 384
Format 48
Format 64
Unit Size:
1mL
This item is not returnable. View return policy

Product Code. 15504364

Brand: ABI Applied Biosystems 4478260

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Custom PreAmp Pools

Custom miRNA array designs can hold from 12 to 380 assays, and 1 to 8 samples on each array design. Ten formats are offered, available only with microfluidics card design. Use Custom PreAmp pools for samples with limited miRNA amounts.
TRUSTED_SUSTAINABILITY

Specifications

Type Custom TaqMan™ miRNA Pre-Amp Pool
Format Format 384
Content And Storage Room Temperature
No. of Samples 1 to 8
pH 8
Quantity 1 mL
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.