Learn More
Bode Chemie™ Cutasept™ F
Product Code. 10460891
Change view
Click to view available options
Volume (Metric):
1 L
Unit Size:
1 L
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Volume (Metric) Unit Size
10460891 1 L 1 L
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Product Code. 10460891 Supplier Bode Chemie™ Supplier No. 9803630
  • ONLINE EXCLUSIVE PRICE
    € 24.15
    € 10.26 / 1 L
    Save € 13.89
    57% Off
 Restricted Item
Estimated Shipment: 30-10-2026
Add to basket
Add to basket
missing translation for 'validMainframeMsg'
Cutasept™ F is a colorless propanol-based skin antiseptic. For use before injections, punctures and surgical interventions with a quick and comprehensive effect.
  • Colorless
  • Works quickly and comprehensively
  • 24 hour long-term effect
  • Excellent skin tolerance
  • Practical application thanks to the overhead spray system
  • Required spectrum of activity SARS-CoV-2: limited virucidal activity
Specifications
Product Type Antiseptic
Color Colorless
For Use With (Application) Before Injections, Punctures and Surgical Interventions, For Diabetics As Part of Blood Sugar Measurement and Insulin Administration
Volume (Metric) 1 L