Learn More
B Braun™ Cutfix Stainless Steel Scalpels
Use B Braun™ Cutfix Sterile Stainless Steel Scalpels, with their double blade support, for precise cutting as your needs. This scalpel has a plastic handle.
Brand: B Braun™ 5518091
Additional Details : Weight : 0.12600kg
Description
- Disposable
- Sterile
- Stainless steel
- Two-sided blade support
Specifications
Stainless Steel | |
# 24 | |
Sterile |
Dental Surgery | |
Yes | |
Plastic |