Learn More
Agilent BioTek Cuvette Adapter
Shop All Agilent BioTek Products
Click to view available options
For Use With (Equipment):
Agilent BioTek Microplate Washers and Readers
Unit Size:
Each
Description
- With this adapter up to 4 standard cuvettes can be placed horizontally and read directly in the plate readers - care must be taken that the cuvette is stoppered or sealed
- The adapter has 4 slots for cuvettes, with aperatures designed for several reading position in a standard microplate footprint
Specifications
Specifications
| Product Type | Cuvette Adapter |
| For Use With (Equipment) | Agilent BioTek Microplate Washers and Readers |
| For Use With (Application) | Designed for Several Reading Positions in a Standard Microplate Footprint |
| Quantity | 1 Pk. |