Learn More
Hach Lange™ Cuvette Tests
For use with water analysis, these exemplary cuvette tests have ISO 6878-1-1986 and DIN 38405 D11-4 certifications. They utilize the Phosphormolybdenum blue testing method for the DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments.
Brand: Hach Lange™ LCK350
Additional Details : Weight : 0.50000kg Transport : UN number : 3316 Chem class : 9 Pack group : II
Description
- Phosphate (ortho/total) cuvette test 2.0 -20.0 mg/L PO4-P
- ISO 6878-1-1986, DIN 38405 D11-4 certifications
- 2 to 20 and 6 to 60mg/L range
- For use with Water analysis
- For use with DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 instruments
- 25 Tests
- Shelf life of 24 Months
- Phosphormolybdenum blue testing method
Specifications
ISO 6878-1-1986, DIN 38405 D11-4 | |
DR3900, DR6000, DR1900, DR2800, DR3800, DR5000 | |
1 Pack | |
15°C to 25°C | |
Phosphate Ortho/Total Cuvette Test |
Water analysis | |
25 | |
2 to 20 and 6 to 60mg/L | |
Phosphormolybdenum blue |
Safety and Handling
- Phosphat/Phosphate, DosiCapZip
Signal Word
- Danger
Hazard Category
- Acute toxicity Category 4
- Serious eye damage/eye irritation Category 2
- Respiratory sensitiser Category 1
- Skin sensitiser Category 1
- Specific target organ toxicity Category 3
- Substances and mixtures corrosive to metals Category 1
- Skin corrosion/irritation Category 1 A
Hazard Statement
- H302-Harmful if swallowed.
- H334-May cause allergy or asthma symptoms or breathing difficulties if inhaled.
- H335-May cause respiratory irritation.
- H319-Causes serious eye irritation.
- H315-Causes skin irritation.
- H317-May cause an allergic skin reaction.
- H290-May be corrosive to metals.
- H314-Causes severe skin burns and eye damage.
Precautionary Statement
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P304+P340-IF INHALED: Remove person to fresh air and keep comfortable for breathing.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P337+P313-If eye irritation persists: Get medical advice/attention.
- P342+P311-If experiencing respiratory symptoms: Call a POISON CENTER/doctor/ .
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P301+P330+P331-IF SWALLOWED: rinse mouth. Do NOT induce vomiting.
- P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
- P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
- P310-Immediately call a POISON CENTER/doctor/.
Supplemental information
- MIXTURE LIST-Contains : Sodium peroxidisulfate, Sodium metaborate, Lithium sulfate monohydrate, Sulphuric acid, Ammonium hep