Learn More
Camping Gaz™ CV470 Plus Valve Cartridge
Click to view available options
Capacity (Metric):
450 g
Unit Size:
470g
Specifications
Specifications
| Product Type | Gas Valve Cartridge |
| Capacity (Metric) | 450 g |
| Diameter (Metric) | 110 mm |
| Height (Metric) | 140 mm |
| Includes | Butane/Propane Mix (80/20) |
Safety and Handling
Product Identifier
- Butan
- Danger
- Flammable gas.
- H220-Extremely flammable gas.
- P102-Keep out of reach of children.
- P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
- P377-Leaking gas fire: Do not extinguish, unless leak can be stopped safely.
- P381-Eliminate all ignition sources if safe to do so.
- P403-Store in a well-ventilated place.
- MIXTURE LIST-Contains : Hydrocarbures richesen C3-C4 Gaz de pétrole