Learn More
Macherey-Nagel™ CW Deactivated Capillary Column
Brand: Macherey-Nagel™ 723562.25
Additional Details : Weight : 0.01000kg
Spezifikation
0.53 mm | |
0.8 mm | |
250°C | |
1 Each |
Deactivated Capillary Column | |
Preparation of Capillary Columns Analysis | |
25 m |