missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ CWC15 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals™ NBP3-30399
This item is not returnable.
View return policy
Description
CWC15 Polyclonal antibody specifically detects CWC15 in Human samples. It is validated for Immunoprecipitation,Western Blot
Specifications
| CWC15 | |
| Polyclonal | |
| Tris-citrate/phosphate pH 7 -8. | |
| Rabbit | |
| Affinity purified | |
| RUO | |
| 51503 | |
| Store at 4°C. Do not freeze. | |
| IgG |
| Immunoprecipitation, Western Blot | |
| Unconjugated | |
| C11orf5ORF5, CWC15 homolog, CWC15 homolog (S. cerevisiae), CWC15 spliceosome-associated protein homolog (S. cerevisiae), Cwf15, HSPC148, protein CWC15 homolog | |
| Between 75 and 125 | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction