missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CWC15 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA558962
This item is not returnable.
View return policy
Description
Immunogen sequence: RGGRGKGEGD LSQLSKQYSS RDLPSHTKIK YRQTTQDAPE EVRNRDFRRE LEERERAAAR EKNRDRPTRE HTTSSSVS Highest antigen sequence identity to the following orthologs: Mouse - 99%, Rat - 100%.
Cwc15 gene ontology annotations related to this gene include mRNA splicing, via spliceosome.
Specifications
| CWC15 | |
| Polyclonal | |
| Unconjugated | |
| Cwc15 | |
| 0610040D20Rik; 2900052N06Rik; AD002; AD-002; C11orf5; CWC15; CWC15 homolog (S. cerevisiae); CWC15 spliceosome-associated protein; CWC15 spliceosome-associated protein homolog; CWC15 spliceosome-associated protein homolog (S. cerevisiae); Cwf15; Ed1; Embryonic development factor 1; HSPC148; mED1; nucleoporin; ORF5; protein CWC15 homolog; putative protein CWC15 like protein; spliceosome-associated protein CWC15 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 51503 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9P013 | |
| Cwc15 | |
| Recombinant protein corresponding to Human CWC15. Recombinant protein control fragment (Product #RP-97379). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction