Learn More
Carbolite™ CWF 12/5 Chamber Furnaces
Click to view available options
Dimensions (D x W x H) Interior:
10 x 5.5 x 5.25in.
5.25 x 5.5 x 10in.
Unit Size:
Each
Specifications
Specifications
| Capacity (Metric) | 5 L |
| Temperature (Metric) | 1200°C |
| Type | Chamber Furnace |
| Includes | Programmable Temperature Controller |
| Dimensions (D x W x H) Interior | 10 x 5.5 x 5.25in. |
| Width (Metric) | 374.65 mm |
| Width (Metric) Interior | 139.7 mm |
| Height (Metric) | 584.2 mm |
| Height (Metric) Interior | 133.35 mm |
| Depth (Metric) | 482.6 mm |
| Show More |