missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CWF19L2 Antibody, DyLight 594, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP1-78215DL594
This item is not returnable.
View return policy
Description
CWF19L2 Polyclonal antibody specifically detects CWF19L2 in Human samples. It is validated for Western Blot
Specifications
| CWF19L2 | |
| Polyclonal | |
| Western Blot | |
| CWF19-like 2, cell cycle control (S. pombe), CWF19-like protein 2, FLJ32343 | |
| The immunogen for this product maps to a region between residue 350 and 400 of human CWF19-like 2 using the numbering given in entry NP_689647.2 (GeneID 143884). | |
| 0.1 mL | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| DyLight 594 | |
| 50mM Sodium Borate | |
| Rabbit | |
| Immunogen affinity purified | |
| RUO | |
| 143884 | |
| Store at 4C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction