missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ CWH43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA569647
This item is not returnable.
View return policy
Description
This target displays homology in the following species: Cow: 100%; Dog: 100%; Guinea Pig: 86%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 93%; Rat: 100%.
CWH43 is a protein coding gene.
Specifications
| CWH43 | |
| Polyclonal | |
| Unconjugated | |
| CWH43 | |
| C130090K23Rik; cell wall biogenesis 43 C-terminal homolog; cell wall biogenesis 43 C-terminal homolog (S. cerevisiae); cell wall biogenesis protein 43 C-terminal homolog; CWH43; CWH43-C; imds-60; PGAP2-interacting protein; PGAP2IP; RGD1310958 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 80157 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9H720 | |
| CWH43 | |
| synthetic peptide directed towards the C-terminal region of human CWH43. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction