missing translation for 'onlineSavingsMsg'
Learn More

Tocris Bioscience™ CX08005
GREENER_CHOICE

Product Code. 18785369
Click to view available options
Quantity:
5 mg
25 mg
Unit Size:
25mg
5mg
This item is not returnable. View return policy

Product Code. 18785369

Brand: Tocris Bioscience™ 6144/25

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Competitive PTP1B inhibitor

CX08005 is a competitive PTP1B inhibitor (IC50 = 0.781 μM). Binds catalytic P-loop of enzyme. Improves insulin sensitivity in obese mice in the glucose tolerance test and the homeostasis model assessment of insulin resistance index (HOMA-IR) . Enhances insulin-induced glucose uptake in adipocytes and myotubes in vitro. Exhibits some selectivity for PTP1B over other protein tyrosine phosphatases, except TCPTP.
TRUSTED_SUSTAINABILITY

Specifications

Chemical Name or Material 2-[[[2-(Tetradecyl)phenyl]amino]carbonyl]benzoic acid
CAS 1256341-22-6
Quantity 25 mg
Molecular Formula C28H39N04
Purity 98%
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.