missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CL1/Fractalkine Antibody (126315), CoraFluor™ 1, Novus Biologicals™
Rat Monoclonal Antibody
Brand: Novus Biologicals FAB571CL1
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Monoclonal antibody specifically detects CX3CL1/Fractalkine in Mouse samples. It is validated for Western Blot,ELISA Standard (Matched Pair),Neutralization,ELISA Capture (Matched Antibody Pair),ELISA Detection (Matched Antibody Pair)
Specifications
| CX3CL1/Fractalkine | |
| Monoclonal | |
| CoraFluor 1 | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| Mouse myeloma cell line NS0-derived recombinant mouse CX3CL1/Fractalkine, S. frugiperda insect ovarian cell line Sf 21-derived recombinant mouse CX3CL1/Fractalkine, and E. coli-derived recombinant mouse CX3CL1/Fractalkine | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 6376 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG2a |
| Western Blot, ELISA Standard (Matched Pair), Neutralization, ELISA Capture (Matched Antibody Pair), ELISA Detection (Matched Antibody Pair) | |
| 126315 | |
| PBS | |
| Rat | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Mouse | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction