missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CL1/Fractalkine Antibody (81506R), Janelia Fluor™ 646, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals FAB3652RJ
This item is not returnable.
View return policy
Description
CX3CL1/Fractalkine Monoclonal antibody specifically detects CX3CL1/Fractalkine in Human samples. It is validated for Neutralization, Flow Cytometry (Intracellular Staining), CyTOF
Specifications
| CX3CL1/Fractalkine | |
| Monoclonal | |
| Janelia Fluor 646 | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified from cell culture supernatant | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Neutralization, Flow Cytometry (Intracellular Staining), CyTOF | |
| 81506R | |
| Neutralization, Intracellular Staining by Flow Cytometry, CyTOF-ready | |
| ABCD-3, C3Xkine, chemokine (C-X3-C motif) ligand 1, CX3C membrane-anchored chemokine, CXC3, CXC3C, FKN, fractalkine, Neurotactin, neurotactin), NTNsmall inducible cytokine subfamily D (Cys-X3-Cys), member 1 (fractalkine, NTTSmall-inducible cytokine D1, SCYD1C-X3-C motif chemokine 1, small inducible cytokine subfamily D (Cys-X3-Cys), member-1 | |
| NS0-derived and E. coli-derived Recombinant human CX3CL1/Fracktalkine, Gln25-Arg339, Accession # P78423 | |
| 0.1 mL | |
| Biologically Active Proteins, Cytokine Research, Immunology, Innate Immunity, Virology Bacteria and Parasites | |
| 6376 | |
| Store at 4°C in the dark. | |
| IgG1 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction