missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CX3CR1 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 13885-1-AP-150UL
This item is not returnable.
View return policy
Description
CX3CR1 is one of the chemokine receptors that are required as coreceptors for HIV infection. The genes encoding human, mouse, and rat CX3CR1 were cloned and designated V28 and CMKBRL1, CX3CR1, and RBS11, respectively. The encoded seven transmembrane protein was recently identified as the receptor for a novel transmembrane molecule, fractalkine, and renamed CX3CR1. Recently, CX3CR1 was found to serve as a coreceptor for HIV-1 and HIV-2 envelope fusion and virus infection, which can be inhibited by fractokine. CX3CR1 mediates leukocyte migration an d adhesion. CX3CR1 is expressed in a variety of human tissues and cell lines.Specifications
| CX3CR1 | |
| Polyclonal | |
| Unconjugated | |
| CX3CR1 | |
| Beta chemokine receptor like 1, C X3 C CKR 1, CCRL1, CMK BRL 1, CMK BRL1, CMKBRL1, CMKDR1, CX3C chemokine receptor 1, CX3CR1, Fractalkine receptor, G protein coupled receptor 13, GPR13, GPRV28, V28 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 13051, 1524, 171056 | |
| -20°C | |
| Liquid |
| Flow Cytometry, Immunohistochemistry (Paraffin), Immunoprecipitation, Western Blot | |
| 0.35 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| P35411, P49238, Q9Z0D9 | |
| CX3CR1 | |
| CX3CR1 Fusion Protein Ag4149 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction