missing translation for 'onlineSavingsMsg'
Learn More

CX3CR1 Rat anti-Human , BB515, Clone: 2A9-1, BD Biosciences™

Rat Monoclonal Antibody

€ 169.38 - € 455.00

Specifications

Antigen CX3CR1
Clone 2A9-1
Applications Flow Cytometry
Classification Monoclonal
Conjugate Brilliant Blue 515 (BB515)
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17237494
View Documents
BD Biosciences
565902
25 Tests
ONLINE EXCLUSIVE PRICE
€ 171.00
€ 169.38 / 25 tests
Save € 1.62
0.95% Off
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17247494
View Documents
BD Biosciences
565903
100 Tests
€ 455.00
100 tests
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
Description

Description

CX3CR1 Monoclonal antibody specifically detects CX3CR1 in Human samples. It is validated for Flow Cytometry
Specifications

Specifications

CX3CR1
Flow Cytometry
Brilliant Blue 515 (BB515)
RUO
Aqueous buffered solution containing ≤0.09% sodium azide.
Human CX3CR1 Recombinant Protein
Primary
2A9-1
Monoclonal
Rat
Human
CCRL1; CMKBRL1; CMKDR1; GPR13; GPRV28; V28; Fractalkine Receptor
IgG2b κ
Store undiluted at 4°C and protected from prolonged exposure to light. Do not freeze. The monoclonal antibody was purified from tissue culture supernatant or ascites by affinity chromatography. The antibody was conjugated with BD Horizon™ BB515 under optimum conditions and unconjugated antibody was removed.
Videos
SDS
Documents

Documents

Product Certifications
Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.