missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXADR Antibody (020), CoraFluor™ 1, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals (Bio-Techne) NBP2-89642CL1
This item is not returnable.
View return policy
Description
CXADR Monoclonal antibody specifically detects CXADR in Human samples. It is validated for ELISA
Specifications
| CXADR | |
| Monoclonal | |
| CoraFluor 1 | |
| CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human CXADR (Accession#: NP_001329.1; Met1-Gly237). | |
| 0.1 mL | |
| Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
| 1525 | |
| Store at 4°C in the dark. Do not freeze. | |
| IgG |
| ELISA | |
| 020 | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction