missing translation for 'onlineSavingsMsg'
Learn More
Learn More
CXADR Rabbit anti-Human, Clone: 23, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-89643-50UL
This item is not returnable.
View return policy
Description
CXADR Monoclonal specifically detects CXADR in Human samples. It is validated for Sandwich ELISA Capture.
Specifications
| CXADR | |
| Monoclonal | |
| Unconjugated | |
| 0.2 um filtered solution in PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| ELISA | |
| 23 | |
| Sandwich ELISA Capture 1:250-1:2000 | |
| CAR10Coxsackievirus B-adenovirus receptor, CAR4/6, coxsackie virus and adenovirus receptor, coxsackie virus B receptor, coxsackievirus and adenovirus receptor, CVB3 binding protein, CVB3-binding protein, hCAR, HCVADR | |
| Recombinant Human CXADR Protein | |
| 50 μg | |
| Cell Biology, Stem Cell Markers, Virology Bacteria and Parasites | |
| 1525 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction